Wasser, HyClone™ HyPure USP, WFI (Wasser für Injektionszwecke)-Qualität

Lieferant: HyClone products (Cytiva)

SH31191.02 SH31191.01 SH31191.03 SH31191.LS SH31191.04
SH31191.02EA 31.7 EUR
SH31191.02 SH31191.01 SH31191.03 SH31191.LS SH31191.04
Wasser, HyClone™ HyPure USP, WFI (Wasser für Injektionszwecke)-Qualität
Wasser

Support cell culture growth and biomanufacturing processes with Water for Injection (WFI) quality water.


  • Meets stringent USP specifications
  • Manufactured in ISO 9001 certified facility
  • 0.1 µm sterile filtered
  • Shelf life 24 months


HyClone HyPure WFI Quality Water undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP). Water for injection (WFI) is produced in ISO 9001 certified facilities to ensure a high quality product.


HyPure WFI Quality Water meets the following criteria: Must be produced from a validated water purification system that meets or exceeds current USP requirements for HyPure WFI Quality Water. Must have a quality assurance monitoring system that ensures continuous compliance and incorporates system test and action limits. Must meet all applicable chemical, purity, and other acceptance criteria as outlined in the current USP for both system and packaged water. Must include a lot-specific CoA for each lot of water produced, listing both acceptance criteria and actual test results.


Bitte beachten Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.

Formel: H₂O
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR