Human Recombinant IL-4 (from E. coli)
Lieferant: Shenandoah Biotechnology
100-09AF-100UG
100-09AF-1MG
100-09AF-20UG
100-09AF-500UG
100-09AF-5UG
SHBT100-09AF-100UGEA
501
EUR
SHBT100-09AF-100UG
SHBT100-09AF-1MG
SHBT100-09AF-20UG
SHBT100-09AF-500UG
SHBT100-09AF-5UG
Human Recombinant IL-4 (from E. coli)
Proteine und Peptide
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
- High quality
- Low endotoxin
- Affordable
Bitte beachten This product is for research use only and is not for use in humans.