Human Recombinant IL-4 (from E. coli)
Lieferant: Shenandoah Biotechnology
100-09AF-100UG 100-09AF-1MG 100-09AF-20UG 100-09AF-500UG 100-09AF-5UG
SHBT100-09AF-100UGEA 501 EUR
SHBT100-09AF-100UG SHBT100-09AF-1MG SHBT100-09AF-20UG SHBT100-09AF-500UG SHBT100-09AF-5UG
Human Recombinant IL-4 (from E. coli)
Proteine und Peptide
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
  • High quality
  • Low endotoxin
  • Affordable
Bitte beachten This product is for research use only and is not for use in humans.