Human Recombinant Omentin (from E. coli)
Lieferant: Shenandoah Biotechnology
100-140-100UG 100-140-10UG 100-140-1MG 100-140-500UG
SHBT100-140-100UGEA 699 EUR
SHBT100-140-100UG SHBT100-140-10UG SHBT100-140-1MG SHBT100-140-500UG
Human Recombinant Omentin (from E. coli)
Proteine und Peptide
Omentin is an adipokine that is produced and secreted by the small intestine, visceral adipose tissue, perivascular adipose tissue, and epicardial adipose tissue. Omentin enhances insulin-stimulated glucose uptake in adipocytes and is a link between obesity and Type 2 Diabetes. Omentin also functions as a vasodilator and plays a protective role during coronary atherosclerosis and hypertension.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Bitte beachten For research use only.