Human Recombinant MIF (from E. coli)
Lieferant: Shenandoah Biotechnology
100-153-100UG 100-153-1MG 100-153-25UG 100-153-500UG 100-153-5UG
SHBT100-153-100UGEA 453 EUR
SHBT100-153-100UG SHBT100-153-1MG SHBT100-153-25UG SHBT100-153-500UG SHBT100-153-5UG
Human Recombinant MIF (from E. coli)
Proteine und Peptide
Macrophage migration inhibitory factor (MIF) is a pro-inflammatory lymphokine that functions during cell-mediated immmunity. MIF promotes fibroblast migration by inducing interleukin 1 (IL-1), interleukin 8 (IL-8), and matrix metalloproteinase (MMP) expression. In interferon-gamma-activated macrophages, MIF stimulates nitric oxide (NO) production and tumor necrosis factor-alpha (TNF-α) secretion.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Lieferumfang: Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Bitte beachten For research use only.