Human Recombinant IGF-I (from E. coli)
Lieferant: Shenandoah Biotechnology
100-34-100UG 100-34-20UG 100-34-5UG
SHBT100-34-100UGEA 181 EUR
SHBT100-34-100UG SHBT100-34-20UG SHBT100-34-5UG
Human Recombinant IGF-I (from E. coli)
Proteine und Peptide
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death. Mature human IGF-I is 100% homologous to bovine and porcine IGF-1 proteins.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.  
Bitte beachten For research use only.