Human Recombinant PTN (Animal free) (from E. coli)
Lieferant: Shenandoah Biotechnology
100-55AF-100UG 100-55AF-1MG 100-55AF-20UG 100-55AF-500UG 100-55AF-5UG
SHBT100-55AF-100UGEA 501 EUR
SHBT100-55AF-100UG SHBT100-55AF-1MG SHBT100-55AF-20UG SHBT100-55AF-500UG SHBT100-55AF-5UG
Human Recombinant PTN (Animal free) (from E. coli)
Proteine und Peptide
Connective tissue growth factor (CTGF) is a mitogen that is secreted by vascular endothelial cells in response to basic fibroblast growth factor (bFGF) or vascular endothelial growth factor (VEGF). CTGF promotes cell growth, migration, adhesion, and survival of endothelial cells.
  • High quality
  • Low endotoxin
  • Affordable
CTGF is also important during osteogenesis, chondrogenesis, and skeletogenesis. CTGF has an insulin-like growth factor binding protein (IGFBP) domain, a thrombospondin type 1 repeat (TSR) domain, and a C-terminal cysteine knot motif.
It is also binds the receptor protein tyrosine phosphatase type Z (PTPRZ), syndecan-3, and anaplastic lymphoma kinase (ALK) receptors.
Lieferumfang: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Bitte beachten For research use only.