Human Recombinant IL-19 (from E. coli)
Lieferant: Shenandoah Biotechnology
100-88-100UG 100-88-10UG 100-88-1MG 100-88-500UG
SHBT100-88-100UGEA 699 EUR
SHBT100-88-100UG SHBT100-88-10UG SHBT100-88-1MG SHBT100-88-500UG
Human Recombinant IL-19 (from E. coli)
Proteine und Peptide
Interleukin-19 (IL-19) is a member of the interleukin 10 (IL-10) cytokine family and is produced by B cells and monocytes. IL-19 binds the interleukin 20 receptor complex (IL-20R) to activate STAT3 signaling. IL-19 induces interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-a) expression in monocytes, and promotes T helper 2 (Th2) cell-mediated immune responses. IL-19 production is up-regulated in resting monocytes following granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS) stimulation.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Bitte beachten For research use only.