Mouse Recombinant IGF-I (from E. coli)
Lieferant: Shenandoah Biotechnology
200-05-100UG 200-05-10UG 200-05-1MG 200-05-500UG 200-05-50UG
SHBT200-05-100UGEA 327 EUR
SHBT200-05-100UG SHBT200-05-10UG SHBT200-05-1MG SHBT200-05-500UG SHBT200-05-50UG
Mouse Recombinant IGF-I (from E. coli)
Proteine und Peptide
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Lieferumfang: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Bitte beachten For research use only.