Macaque Recombinant R EBI3 SUBUNIT (IL-27/IL-35) (from E. coli)
Lieferant: Shenandoah Biotechnology
400-07-20UG 400-07-50UG 400-07-250UG 400-07-500UG 400-07-1MG
SHBT400-07-20UGEA 72.4 EUR
SHBT400-07-20UG SHBT400-07-50UG SHBT400-07-250UG SHBT400-07-500UG SHBT400-07-1MG
Macaque Recombinant R EBI3 SUBUNIT (IL-27/IL-35) (from E. coli)
Proteine und Peptide
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
  • High quality
  • Low endotoxin
  • Affordable
Lieferumfang: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Bitte beachten For research use only.