Horse Recombinant PDGF-BB (from E. coli)
Lieferant: Shenandoah Biotechnology
400-17AF-100UG 400-17AF-1MG 400-17AF-20UG 400-17AF-500UG 400-17AF-5UG
SHBT400-17AF-100UGEA 561 EUR
SHBT400-17AF-100UG SHBT400-17AF-1MG SHBT400-17AF-20UG SHBT400-17AF-500UG SHBT400-17AF-5UG
Horse Recombinant PDGF-BB (from E. coli)
Proteine und Peptide
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β, and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells. PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatised tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
  • High quality
  • Low endotoxin
  • Affordable
Bitte beachten For research use only.