Human Recombinant FLT-3 (CTGrade GMP) (from E. coli)
Lieferant: Shenandoah Biotechnology
IL-6 is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages, and muscle cells. Signals through a receptor complex containing two receptors, IL-6Rα and gp130. Plays an important function in promoting fever and can serve to stimulate an immune response to trauma.
- High quality
- Low endotoxin
- Lot-to-lot consistency
- Animal free
- Manufactured following GMP guidelines
IL-6 is often used for growth of hybridoma cell lines. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
Host cell protein: ≤30 ng/ml
Host cell DNA: ≤0,01 ng/ml
Sterility testing: No growth
Mycoplasma: Negative
Bitte beachten For research use or further manufacturing only.