Mouse Monocyte-Derived Dendritic Cell Differentiation Recombinant Protein Bundle (rm GM-CSF/rm IL-4) (from E. coli)
Lieferant: Shenandoah Biotechnology
PB-500-02
SHBTPB-500-02EA 736 EUR
SHBTPB-500-02
Mouse Monocyte-Derived Dendritic Cell Differentiation Recombinant Protein Bundle (rm GM-CSF/rm IL-4) (from E. coli)
Proteine und Peptide
Cytokines for the differentation of mouse monycyte-derived dendritic cells. Dendritic cells are an important type of immune cell that function to activate T and B cells via their role as an antigen presenting cells. Monocytes isolated from bone marrow can be differentiated to immature dendritic cells (iDCs) through culturing in the presence of interleukin 4 (IL-4) and granulocyte-macrophage colony stimulating factor (GM-CSF). The characteristics of monocyte-derived iDCs are detectable after culture with IL-4/GM-CSF and include: the down regulation of CD14 surface expression, an increase in dextran uptake and an increased response to MIP-1α. Subsequent culturing with pro-inflammatory cytokines (such as TNF-α, IL-1β, and IL-6) can further differentiate the iDCs into mature dendritic cells (mDCs) that have full T cell stimulating capacity. This bundle includes 1x 100 μg of Recombinant Mouse IL-4 and 1x 100 μg of Recombinant Mouse GM-CSF.
  • High Quality
  • Low Endotoxin
  • Affordable
This bundle includes 1×100 μg of Recombinant Mouse IL-4 and 1×100 μg of Recombinant Mouse GM-CSF. Specific concentrations of each chemokine are determined by the end user based on their application and culture conditions.
Lieferumfang: Bundle includes: 100 μg of recombinant mouse IL-4; 100 μg of recombinant mouse GM-CSF.
Bitte beachten For research use only.
Neuer Artikel bei VWR!
Bestellen
TECHNISCHE DATEN
Protein/Peptid-Name Mouse monocyte/DC PB
Environmentally Preferable
Protein/Peptid-Typ Rekombinant
Protein-Funktion Cytokine
Spezies Mouse
Ursprung E. coli
Konjugation Unconjugated
Frei von Proteasen N
Tierfrei N
Frei von Nukleasen N
Frei von Trägern Y
Frei von Endotoxinen N
Endotoxinspiegel Low
Sorte RUO
Lagerbedingungen At or below –20 °C
Transporttemperatur RT