Human Recombinant Human Th17 Cell Polarizing PB (from E. coli)
Lieferant: Shenandoah Biotechnology
PB-500-11
SHBTPB-500-11EA 218 EUR
SHBTPB-500-11
Human Recombinant Human Th17 Cell Polarizing PB (from E. coli)
Proteine und Peptide
Cytokines for polarising human T cells into Th17 cells. T helper 17 cells (Th17) are a distinct type of T cell subset that are thought to be important in immunity against microbes at the epitilial and mucosal interfaces of the body. Disruption of the regular function or balance of Th17 cells is thought to be involved in several autoimmune diseases and/or opportunistic infections. Th17 cells are characterised by their ability to produce IL-17, IL-22 and IL-21. Th17 cells can be cultured from naive CD4+ T cells by culturing them in the presence of anti-CD3 and anti-CD28 stimulation with recombinant IL-6 and recombinant TGF-β 1.
  • High quality
  • Low endotoxin
  • Affordable
Bitte beachten For research use only.