Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Lieferant: Shenandoah Biotechnology
PB-500-20
SHBTPB-500-20EA 1450 EUR
SHBTPB-500-20
Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Proteine und Peptide
Cytokines for cell culture of mouse glioma cells. A glioma is a generic term used to describe tumors that arise from the glia tissue of the brain that supports and surrounds neurons. Glioma tumor cells can be successfully cultured ex vivo with EGF, FGF-basic (or FGF-2), and PDGFs (AA and BB).
  • High quality
  • Low endotoxin
  • Affordable
Lieferumfang: 2×100 μg of Recombinant mouse EGF, 200 μg of Recombinant mouse FGF-basic/FGF-2, 100 μg of Recombinant mouse PDGF-AA and 100 μg of Recombinant mouse PDGF-BB.
Bitte beachten For research use only.