Novec™ 7100 engineered fluid

Lieferant: SIGMA ALDRICH MICROSCOPY

SHH0002-500ML
SHH0002-500MLEA 187 EUR
SHH0002-500ML
Novec™ 7100 engineered fluid
Novec™ 7100 engineered fluid

Novec™ 7100 fluid consists of two inseparable isomers ((CF₃)₂CFCF₂OCH₃) and CF₃CF₂CF₂CF₂OCH₃) with essentially identical properties. Ideal for vapor degreasing applications as a neat (pure), azeotropic component and co-solvent parts cleaner.


  • Non-flammable and non-conductive
  • Compatible with plastics
  • Low toxicity and surface tension
  • Low global warming potential (GWP) and zero ozone depletion potential
  • Histology Bath Fluid


Used as a heat transfer fluid, cleaning and rinsing agent, spray contact cleaner, cryofreezing solution, lubricant carrier, dielectric test media and replacement for CFC, HCFC, HFC and PFC.


Novec is a trademark of 3M Company

Formel: C₅H₃OF₉
Molecular Weight: 250.06 g/mol
MDL: MFCD28505595
CAS-Nummer: 163702-08-7;163702-07-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b336d254c14378347175070589e691c3d85c5da2&vid=c54b78ea5cdf160ffd6337c908dc4bff68a63bda&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR