Mini hand press, MHP-1
Lieferant: SHIMADZU
200-66747-91
SHIM200-66747-91EA 2020 EUR
SHIM200-66747-91
Mini hand press, MHP-1
Pressen
This compact mini hand press is a used to produce KBr pellets.
  • Pellet produced in the frame directly measured using the dedicated holder
  • For regular Ø 13 mm pellets, evacuable die, hydraulic press and vacuum pump are used
  • No dies or vacuum pump are required
Bestellinformation: Includes mini hand press, pellet holder, upper and lower cylinder, briquetting frame (Ø 4 mm), pellet remover and pellet remover base.