Software, IRsolution Agent
Lieferant: SHIMADZU
206-21600-92
SHIM206-21600-92EA 899 EUR
SHIM206-21600-92
Software, IRsolution Agent
Software Labor-Software
IRsolution Agent is a tool for managing data from IRsolution results in a database. This allows managing analytical data records over their entire life cycle, from creation (measurement), review, approval, storage, viewing, backup, and disposal.
  • Provides security management and electronic signature functinos for data stored and recorded in a database
  • Measurement data is automatically saved in a database to allow easy searching of target data at a later time
  • Spectral waveform data, peak detection tables and other information stored in the database can be viewed directly
Standards: Compliant with FDA 21 CFR Part 11 (electronic records and electronic signatures).