EXP®2 TI-LOK™ All-in-One PEEK Ferrule, ¹/₁₆"
Lieferant: SHIMADZU
980-33016
SHIM980-33016EA 294 EUR
SHIM980-33016
EXP®2 TI-LOK™ All-in-One PEEK Ferrule, ¹/₁₆"
Fittings und Verbinder
High-Performance fittings for liquid chromatography simple, hand-tightened connections for all LC applications achieve zero dead volume with Titanium and PEEK technology high-pressure, reusable connections for HPLC and UHPLC.
  • Ideal for HPLC and UHPLC connections in tight spaces (injection valves, columns and more)
  • Zero dead volume - Unique design for tight connections without tubing deformation
  • Reusable
  • Compatible tubing
  • high pressure resistance, compact, tool-free
The secret lies in a combination of innovative materials and design. The fittingsMore Product Information