cis-4-Hepten-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: cis-1-Hydroxy-4-hepten, cis-Hept-4-en-1-ol

M-Clarity™ Qualitätsstufe = MQ100

00496-5ML
SIAL00496-5MLEA 39.9 EUR
SIAL00496-5ML
cis-4-Hepten-1-ol, Sigma-Aldrich®
cis-4-Hepten-1-ol
Formel: C₇H₁₄O
Molecular Weight: 114,19 g/mol
MDL: MFCD00015575
CAS-Nummer: 6191-71-5
EINECS: 228-238-2
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR