Rebaudioside A ≥96% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl (4R,4aS,6aR,9S,11aR,11bS)-9-(((2S,3R,4S,5R,6R)-5-hydroxy-6-(hydroxymethyl)-3,4-bis(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate

01432-10G
SIAL01432-10GEA 123 EUR
SIAL01432-10G
Rebaudioside A ≥96% (durch HPLC), Sigma-Aldrich®
Rebaudioside A

Rebaudioside A is a glucosylated steviol glycoside studied and used as a non-glycemic food sweetener.

Formel: C₄₄H₇₀O₂₃
Molecular Weight: 967.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02183463
CAS-Nummer: 58543-16-1

Bestellen

Spezifikation Testergebnisse

Assay (HPLC) ≥96%
Optical Activity [α]/D –33.0±3.0°, c=1% in H₂O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR