Acridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

01640-10G
SIAL01640-10GEA 147 EUR
SIAL01640-10G
Acridin, Sigma-Aldrich®
Acridine
Formel: C₁₃H₉N
Molecular Weight: 179,22 g/mol
MDL: MFCD00005025
CAS-Nummer: 260-94-6
EINECS: 205-971-6
UN: 2713
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR