Adenin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Purin-6-amin Hydrochlorid, 1H-Adenin Hydrochlorid, 9H-Purin-6-amin Hydrochlorid, 6-Aminopurin Hydrochlorid
M-Clarity™ Qualitätsstufe = MQ200
01840-25G
SIAL01840-25GEA
48.1
EUR
SIAL01840-25G
Adenin Hydrochlorid, Sigma-Aldrich®
Adenin Hydrochlorid
Formel:
C₅H₅N₅·HCl Molecular Weight: 171,59 g/mol |
MDL:
MFCD00150864 CAS-Nummer: 2922-28-3 EINECS: 220-867-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...