Bis(2-ethylhexyl)adipat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate

M-Clarity™ Qualitätsstufe = MQ100

02140-1L
SIAL02140-1LEA 42 EUR
SIAL02140-1L
Bis(2-ethylhexyl)adipat, Sigma-Aldrich®
Bis(2-ethylhexyl)adipat
Formel: C₂₂H₄₂O₄
Molecular Weight: 370.57 g/mol
MDL: MFCD00009496
CAS-Nummer: 103-23-1
EINECS: 203-090-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR