1,1,2-Trimethyl-1H-benz[e]indol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1,2-Trimethyl-1H-benzo[e]indole

M-Clarity™ Qualitätsstufe = MQ200

03024-25G-F
SIAL03024-25G-FEA 95.3 EUR
SIAL03024-25G-F
1,1,2-Trimethyl-1H-benz[e]indol, Sigma-Aldrich®
1,1,2-Trimethyl-1H-benz[e]indol
Formel: C₁₅H₁₅N
Molecular Weight: 209,29 g/mol
MDL: MFCD00082627
CAS-Nummer: 41532-84-7
EINECS: 255-429-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR