N-Ethyldiisopropylamin 2 mol/l in N-Methyl-2-pyrrolidon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
03439-250ML
SIAL03439-250MLEA
95.6
EUR
SIAL03439-250ML
N-Ethyldiisopropylamin 2 mol/l in N-Methyl-2-pyrrolidon, Sigma-Aldrich®
N-Ethyldiisopropylamin
Formel:
C₈H₁₉N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008868 CAS-Nummer: 7087-68-5 EINECS: 230-392-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...