Dimethyl-5-bromisophthalat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-1,3-benzoldicarbonsäuredimethylester, 5-Bromisophthalsäuredimethylester

M-Clarity™ Qualitätsstufe = MQ100

04077-5G
SIAL04077-5GEA 301 EUR
SIAL04077-5G
Dimethyl-5-bromisophthalat, Sigma-Aldrich®
Dimethyl-5-bromisophthalat
Formel: C₁₀H₉BrO₄
Molecular Weight: 273,08 g/mol
MDL: MFCD00078709
CAS-Nummer: 51760-21-5
EINECS: 257-386-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR