1-(3-Cyanpropyl)-3-methylimidazoliumchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

04124-25G-F
SIAL04124-25G-FEA 893 EUR
SIAL04124-25G-F
1-(3-Cyanpropyl)-3-methylimidazoliumchlorid, Sigma-Aldrich®
1-(3-Cyanopropyl)-3-methylimidazolium chloride
Formel: C₈H₁₂ClN₃
CAS-Nummer: 683224-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR