(S)-3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-3-(3,4-dimethoxyphenyl)propanoic acid, (S)-Boc-3,4-dimethoxy-β-Phe-OH

M-Clarity™ Qualitätsstufe = MQ100

04542-500MG-F
SIAL04542-500MG-FEA 365 EUR
SIAL04542-500MG-F
(S)-3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionsäure
Formel: C₁₆H₂₃NO₆
MDL: MFCD03427907
CAS-Nummer: 499995-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR