(S)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-3-(4-fluorophenyl)propanoic acid

M-Clarity™ Qualitätsstufe = MQ100

05102-500MG-F
SIAL05102-500MG-FEA 123 EUR
SIAL05102-500MG-F
(S)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure
Formel: C₁₄H₁₈FNO₄
Molecular Weight: 283,2954 g/mol
MDL: MFCD03427919
CAS-Nummer: 479064-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR