3-(2-Aminoethylamino)propylmethyldimethoxysilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(2-Aminoethylamino)propyldimethoxymethylsilane

M-Clarity™ Qualitätsstufe = MQ100

06667-25ML
SIAL06667-25MLEA 77.8 EUR
SIAL06667-25ML
3-(2-Aminoethylamino)propylmethyldimethoxysilan, Sigma-Aldrich®
3-(2-Aminoethylamino)propylmethyldimethoxysilan
Formel: C₈H₂₂N₂O₂Si
Molecular Weight: 206,36 g/mol
MDL: MFCD00054776
CAS-Nummer: 3069-29-2
EINECS: 221-336-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR