(1S,2S)-trans-2-Aminocyclopentanol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-(1S,2S)-2-Aminocyclopentanol Hydrochloride, (1S,2S)-2-Aminocyclopentanol hydrochloride, (1S,2S)-trans-2-Aminocyclopentanol hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

06782-1G-F
SIAL06782-1G-FEA 250 EUR
SIAL06782-1G-F
(1S,2S)-trans-2-Aminocyclopentanol Hydrochlorid, Sigma-Aldrich®
(1S,2S)-trans-2-Aminocyclopentanol Hydrochlorid
Formel: C₅H₁₂ClNO
Molecular Weight: 137.6079 g/mol
MDL: MFCD07370092
CAS-Nummer: 68327-04-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR