4-Aminobenzamidin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

06880-5G
SIAL06880-5GEA 224 EUR
SIAL06880-5G
4-Aminobenzamidin Dihydrochlorid, Sigma-Aldrich®
4-Aminobenzamidin Dihydrochlorid
Formel: C₇H₁₁Cl₂N₃
Schmelzpunkt: 290…291 °C
MDL: MFCD00013001
CAS-Nummer: 2498-50-2
EINECS: 219-692-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR