(S)-3-(Boc-amino)-3-(2-methylphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-3-(o-tolyl)propanoic acid

M-Clarity™ Qualitätsstufe = MQ100

06939-500MG-F
SIAL06939-500MG-FEA 364 EUR
SIAL06939-500MG-F
(S)-3-(Boc-amino)-3-(2-methylphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(2-methylphenyl)propionsäure
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,3300 g/mol
MDL: MFCD03427895
CAS-Nummer: 499995-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR