(4-tert-Butylphenoxy)essigsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

0710000-25G
SIAL0710000-25GEA 69.6 EUR
SIAL0710000-25G
(4-tert-Butylphenoxy)essigsäure, Sigma-Aldrich®
(4-tert-Butylphenoxy)essigsäure
Formel: (C₆H₄(C(CH₃)₃))CH₂COOH
MDL: MFCD00021758
CAS-Nummer: 1798-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR