5-Nitro-2-thiophencarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Nitrothiophene-2-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

07517-1G-F
SIAL07517-1G-FEA 51.8 EUR
SIAL07517-1G-F
5-Nitro-2-thiophencarbonitril, Sigma-Aldrich®
5-Nitro-2-thiophencarbonitril
Formel: C₅H₂N₂O₂S
Molecular Weight: 154,15 g/mol
MDL: MFCD00052401
CAS-Nummer: 16689-02-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR