Synperonic® F 108 surfactant, non-ionic, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Synperonic® F108, PEG-PPG-PEG, Pluronic® F-108

M-Clarity™ Qualitätsstufe = MQ100

07579-250G-F
SIAL07579-250G-FEA 77.9 EUR
SIAL07579-250G-F
Synperonic® F 108 surfactant, non-ionic, Sigma-Aldrich®
Pluronic® F-108 (Synperonic® F 108)

Synperonic F-108 is a polyoxyethylene-polyoxypropylene block copolymer. It has two hydrophilic groups that are attached to a central hydrophobic group that self-assembles in an aqueous solution into clusters known as micelles. This water-soluble surfactant is used as effective emulsification and solubilizing agent in a wide range of applications.


Synperonic is a registered trademark of Croda International PLC

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Molecular Weight: 14600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR