2-(Methylthio)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

07655-1G-F
SIAL07655-1G-FEA 110 EUR
SIAL07655-1G-F
2-(Methylthio)pyridin, Sigma-Aldrich®
2-(Methylthio)pyridine
Formel: C₆H₇NS
MDL: MFCD01691593
CAS-Nummer: 18438-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR