(R)-(-)-3-Amino-1-Boc-piperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-butyl (3R)-3-aminopiperidine-1-carboxylate, (R)-1-Boc-3-Aminopiperidine

M-Clarity™ Qualitätsstufe = MQ100

07858-1G-F
SIAL07858-1G-FEA 267 EUR
SIAL07858-1G-F
(R)-(-)-3-Amino-1-Boc-piperidin, Sigma-Aldrich®
(R)-(-)-3-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
MDL: MFCD03094717
CAS-Nummer: 188111-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR