(R)-3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-(3,4-dimethoxyphenyl)propanoic acid, Boc-D-β-Phe(3,4-DiOMe)-OH

M-Clarity™ Qualitätsstufe = MQ100

08009-500MG-F
SIAL08009-500MG-FEA 340 EUR
SIAL08009-500MG-F
(R)-3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionsäure, Sigma-Aldrich®
(R)-3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionsäure
Formel: C₁₆H₂₃NO₆
Molecular Weight: 325,356 g/mol
MDL: MFCD03427945
CAS-Nummer: 500788-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR