Methylrot 0,1 g/l in Ethanol 57% (v/v), Millipore®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
08714-100ML-F
SIAL08714-100ML-FEA
44.3
EUR
SIAL08714-100ML-F
Methylrot 0,1 g/l in Ethanol 57% (v/v), Millipore®
Methylrot
Formel:
C₁₅H₁₅N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002425 CAS-Nummer: 493-52-7 EINECS: 207-776-1 UN: 1170 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...