(S)-3-(Boc-amino)-3-phenylpropionsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-Boc-beta3-phenylalanine, Boc-D-β-Phe-OH, (S)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid
M-Clarity™ Qualitätsstufe = MQ100
09793-1G
SIAL09793-1GEA
199
EUR
SIAL09793-1G
(S)-3-(Boc-amino)-3-phenylpropionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-phenylpropionsäure
Formel:
C₁₄H₁₉NO₄ |
MDL:
MFCD01860892 CAS-Nummer: 103365-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...