(R)-3-(Boc-amino)-3-phenylpropionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid, (R)-N-BOC-3-Amino-3-phenylpropanoic acid

M-Clarity™ Qualitätsstufe = MQ100

09794-1G
SIAL09794-1GEA 682 EUR
SIAL09794-1G
(R)-3-(Boc-amino)-3-phenylpropionsäure, Sigma-Aldrich®
(R)-3-(Boc-amino)-3-phenylpropionsäure
Formel: C₁₄H₁₉NO₄
Molecular Weight: 265,31 g/mol
MDL: MFCD01320859
CAS-Nummer: 161024-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR