Propargyl-p-toluolsulfonat, Sigma-Aldrich®

Lieferant: Merck

09954-25ML
SIAL09954-25MLEA 218 EUR
SIAL09954-25ML
Propargyl-p-toluolsulfonat, Sigma-Aldrich®
Propargyl-p-toluolsulfonat
Formel: C₁₀H₁₀O₃S
MDL: MFCD00078365
CAS-Nummer: 6165-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR