2,3,5,6-Tetrafluorthiophenol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,3,5,6-Tetrafluorbenzolthiol, 2,3,5,6-Tetrafluorphenylmercaptan, 1-Mercapto-2,3,5,6-tetrafluorbenzol, 2,3,5,6-Tetrafluorbenzol-1-thiol
M-Clarity™ Qualitätsstufe = MQ100
100188-5G
SIAL100188-5GEA
96.7
EUR
SIAL100188-5G
2,3,5,6-Tetrafluorthiophenol, Sigma-Aldrich®
2,3,5,6-Tetrafluorthiophenol
Formel:
C₆H₂F₄S Molecular Weight: 182,14 g/mol Siedepunkt: 152 °C (1013 hPa) |
MDL:
MFCD00004829 CAS-Nummer: 769-40-4 EINECS: 212-210-1 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...