D(+)-Cellobioseoctaacetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
101575-25G
SIAL101575-25GEA
30
EUR
SIAL101575-25G
D(+)-Cellobioseoctaacetat, Sigma-Aldrich®
D(+)-Cellobioseoctaacetat
Formel:
C₂₈H₃₈O₁₉ Molecular Weight: 678,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009600 CAS-Nummer: 5346-90-7 EINECS: 226-304-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...