1-(Cyanacetyl)piperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Oxo-3-(piperidin-1-yl)propanenitrile

M-Clarity™ Qualitätsstufe = MQ100

102180-100G
SIAL102180-100GEA 318 EUR
SIAL102180-100G
1-(Cyanacetyl)piperidin, Sigma-Aldrich®
1-(Cyanacetyl)piperidin
Formel: C₈H₁₂N₂O
Molecular Weight: 152,20 g/mol
MDL: MFCD00006487
CAS-Nummer: 15029-30-8
EINECS: 239-111-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR