N-(3-Hydroxypropyl)phthalimid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(3-Hydroxypropyl)-1,2-benzoldicarbonsäureimid, N-(3-Hydroxypropyl)phthalsäureimid

103063-25G
SIAL103063-25GEA 65.9 EUR
SIAL103063-25G
N-(3-Hydroxypropyl)phthalimid, Sigma-Aldrich®
N-(3-Hydroxypropyl)phthalimid
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,21 g/mol
MDL: MFCD00023097
CAS-Nummer: 883-44-3
EINECS: 212-931-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR