Methyl-3-(methylthio)propionat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

103373-25G
SIAL103373-25GEA 52 EUR
SIAL103373-25G
Methyl-3-(methylthio)propionat, Sigma-Aldrich®
Methyl-3-(methylthio)propionat
Formel: C₅H₁₀O₂S
MDL: MFCD00008565
CAS-Nummer: 13532-18-8
EINECS: 236-883-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR