Cyclopentylmethanol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Cyclopentanmethanol
M-Clarity™ Qualitätsstufe = MQ100
103985-10G
SIAL103985-10GEA
66.8
EUR
SIAL103985-10G
Cyclopentylmethanol, Sigma-Aldrich®
Cyclopentylmethanol
Formel:
C₆H₁₂O Molecular Weight: 100,16 g/mol Siedepunkt: 162 °C (1013 hPa) Dichte: 0,926 g/cm³ (20 °C) |
MDL:
MFCD00001384 CAS-Nummer: 3637-61-4 EINECS: 222-861-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...