N-Phenyl-1-naphthylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Anilinonaphthalene

M-Clarity™ Qualitätsstufe = MQ100

104043-500G
SIAL104043-500GEA 86.2 EUR
SIAL104043-500G
N-Phenyl-1-naphthylamin, Sigma-Aldrich®
N-Phenyl-1-naphthylamin
Formel: C₁₆H₁₃N
Molecular Weight: 219,29 g/mol
MDL: MFCD00003878
CAS-Nummer: 90-30-2
EINECS: 201-983-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR